Invitrogen
Manufacturer Code:PA599449
Catalog # PIPA599449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Cytochrome P450 1A1/2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AHH AHRR aromatic compound inducible aryl hydrocarbon hydroxylase CP11 CYP CYP1 CYP1A1 Cyp1a-1 Cyp1a2 Cyp45c Cypc45c CYPIA1 cytochrome P1-450 cytochrome P1-450 dioxin-inducible cytochrome P450 1A1 cytochrome P450 family 1 subfamily A member 1 cytochrome P450 family 1 subfamily a polypeptide 2 cytochrome P450 form 6 cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c) cytochrome P450 subfamily I polypeptide 1 cytochrome P450 1a1 cytochrome P450 1a1 aromatic compound inducible cytochrome P450 family 1 subfamily A polypeptide 1 cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c) cytochrome P450 subfamily I (aromatic compound-inducible) polypeptide 1 cytochrome P450-C Cytochrome P450MT2 Cytochrome P450MT2A Cytochrome P450MT2B Cytochrome P450-P1 dioxin-inducible flavoprotein-linked monooxygenase Hydroperoxy icosatetraenoate dehydratase microsomal monooxygenase P1-450 P450 P450 form 6 P450-1 P450-C P450DX P450form6 P-450MC P450MT2 P450-P1 xenobiotic monooxygenase; aromatic compound inducible; aryl hydrocarbon hydroxylase; CYP1A1; CYPIA1; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; Cytochrome P450 1A1; cytochrome P450 family 1 subfamily a polypeptide 2; Cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, family 1, subfamily A, polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; Cytochrome P450-C; Cytochrome P450-P1; Cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; dioxin-inducible; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P450form6; P450MT2; xenobiotic monooxygenase
Gene Aliases: AHH; AHRR; CP11; CYP1; Cyp1a-1; CYP1A1; Cyp1a2; Cyp45c; Cypc45c; CYPIA1; P-450MC; P1-450; P450-1; P450-C; P450DX
UniProt ID: (Human) P04798, (Mouse) P00184, (Rat) P00185
Entrez Gene ID: (Human) 1543, (Mouse) 13076, (Rat) 24296
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.