Test
Invitrogen
Manufacturer Code:PA5112867
Catalog # PIPA5112867
Quantity:
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
![]()
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
![]()
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
![]()
Export as a spreadsheet
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer. |
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com.
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase.
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
![]()
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
![]()
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Zebrafish Cytochrome P450 1A1 protein (1-75AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AHH AHRR aromatic compound inducible aryl hydrocarbon hydroxylase CP11 CYP CYP1 CYP1A1 Cyp1a-1 Cyp1a2 Cyp45c Cypc45c CYPIA1 cytochrome P1-450 cytochrome P1-450 dioxin-inducible Cytochrome P450 1A1 cytochrome P450 family 1 subfamily A member 1 cytochrome P450 family 1 subfamily a polypeptide 2 Cytochrome P450 form 6 cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c) cytochrome P450 subfamily I polypeptide 1 cytochrome P450 1a1 cytochrome P450 1a1 aromatic compound inducible cytochrome P450 family 1 subfamily A polypeptide 1 cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c) cytochrome P450 subfamily I (aromatic compound-inducible) polypeptide 1 Cytochrome P450-C Cytochrome P450MT2 Cytochrome P450MT2A Cytochrome P450MT2B Cytochrome P450-P1 dioxin-inducible flavoprotein-linked monooxygenase Hydroperoxy icosatetraenoate dehydratase microsomal monooxygenase P1-450 P450 P450 form 6 P450-1 P450-C P450DX P450form6 P-450MC P450MT2 P450-P1 xenobiotic monooxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.