Invitrogen
Manufacturer Code:PA579124
Catalog # PIPA579124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse Cyp17a1 recombit protein (Position: Q80-R363). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-alpha-hydroxyprogesterone aldolase; CYPXVII; Cytochrome P450 17A1; cytochrome P450, 17; cytochrome P450, 17a1; cytochrome P450, subfamily 17; Cytochrome P450, subfamily XVII; Cytochrome P450-C17; Cytochrome P450c17; P450-C17; P45017alpha; P450c17; steroid 17-alpha hydroxylase; Steroid 17-alpha-hydroxylase/17,20 lyase; Steroid 17-alpha-hydroxylase/1720 lyase; Steroid 17-alpha-monooxygenase
Gene Aliases: Cyp17; Cyp17a1; p450c17
UniProt ID: (Mouse) P27786, (Rat) P11715
Entrez Gene ID: (Mouse) 13074, (Rat) 25146
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.