Invitrogen
Manufacturer Code:PA575966
Catalog # PIPA575966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 82-126 of Human CYP11B1/CYP11B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALDOS; Aldosterone synthase; Aldosterone-synthesizing enzyme; Corticosterone 18-monooxygenase, CYP11B2; CYPXIB1; CYPXIB2; CYPXIB3; Cytochrome P-450Aldo; Cytochrome P-450c11; Cytochrome P-450C18; Cytochrome P450 11B1 mitochondrial CYPXIB1 Cytochrome P-450c11 Cytochrome P450C11 Steroid 11-beta-hydroxylase Cytochrome P450 11B2 mitochondrial Aldosterone synthase ALDOS Aldosterone-synthesizing enzyme CYPXIB2 Cytochrome P-450Aldo Cytochrome P-450C18 Steroid 18-hydroxylase; Cytochrome P450 11B1, mitochondrial; Cytochrome P450 11B2, mitochondrial; cytochrome P450 11B3, mitochondrial; Cytochrome P450 subfamily XIB polypeptide 2 (aldosterone synthase); cytochrome p450 XIB1; cytochrome P450(11 beta)-DS; cytochrome P450, family 11, subfamily B, polypeptide 1; cytochrome P450, family 11, subfamily B, polypeptide 2; cytochrome P450, subfamily 11B, polypeptide 1; cytochrome P450, subfamily 11B, polypeptide 2; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2; Cytochrome P450, subfamily XIB, polypeptide 1 (steroid 11-beta-hydroxylase); Cytochrome P450, subfamily XIB, polypeptide 2 (aldosterone synthase); cytochrome P450-Aldo-1; cytochrome P450-Aldo-2; Cytochrome P450C11; mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2; P450(11 beta)-DS; P450-Aldo-1; P450-Aldo-2; P450C11; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B1; Steroid 11-beta-hydroxylase, CYP11B2; steroid 11-beta-monooxygenase; steroid 11-beta/18-hydroxylase; Steroid 18-hydroxylase; steroid 18-hydroxylase, aldosterone synthase, P450C18, P450aldo; steroid-11-beta-hydroxylase
Gene Aliases: AA389061; ALDOS; Cp11ba; Cp45as; CPN1; CPN2; CYP11B; Cyp11b-1; Cyp11b-2; CYP11B1; CYP11B2; Cyp11b3; CYP11BL; CYPXIB2; FHI; P-450C18; P450aldo; P450C11; P450C18; RATCP11BA; RNCP45AS; S11BH
UniProt ID: (Human) P15538, (Human) P19099, (Mouse) P15539
Entrez Gene ID: (Human) 1584, (Human) 1585, (Mouse) 110115, (Rat) 500892, (Mouse) 13072, (Rat) 24294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.