Novus Biologicals
Manufacturer Code:NBP16888320UL
Catalog # NBP16888320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CYP11B1 (cytochrome P450 family 11 subfamily B polypeptide 1) The peptide sequence was selected from the C terminal of CYP11B1. Peptide sequence ARNPNVQQALRQESLAAAASISEHPQKATTELPLLRAALKETLRLYPVGL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CPN1 CYPXIB1 cytochrome P450 11B1 mitochondrial cytochrome p450 XIB1 cytochrome P450 family 11 subfamily B polypeptide 1 cytochrome P450 subfamily XIB (steroid 11-beta-hydroxylase) polypeptide 1 Cytochrome P-450c11 Cytochrome P450C11 EC 1.14.15 EC 1.14.15.4 FHICYP11B FLJ36771 P450C11DKFZp686B05283 S11BH Steroid 11-beta-hydroxylase steroid 11-beta-monooxygenase; CYPXIB1; Cytochrome P-450c11; Cytochrome P450 11B1, mitochondrial; cytochrome p450 XIB1; cytochrome P450, family 11, subfamily B, polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; Cytochrome P450C11; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B1; steroid 11-beta-monooxygenase
Gene Aliases: CPN1; CYP11B; CYP11B1; FHI; P450C11; S11BH
UniProt ID: (Human) P15538
Entrez Gene ID: (Human) 1584
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.