Proteintech
Manufacturer Code:160111AP150UL
Catalog # 50-173-0640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYFIP1/2 Fusion Protein Ag8749 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYFIP1; CYFIP2; CYFIP2 KIAA1168 p53 inducible protein 121 PIR121; Cytoplasmic FMR1-interacting protein 1; Cytoplasmic FMR1-interacting protein 2; cytoplasmic FMRP interacting protein 1; p140sra-1; p53-inducible protein 121; selective hybridizing clone; Specifically Rac1-associated protein 1; Sra-1
Gene Aliases: 1500004I01Rik; 6430511D02Rik; AA930218; AU022376; CYFIP1; CYFIP2; E030028J09Rik; KIAA0068; KIAA1168; l(7)1Rl; l71Rl; l7Rl1; mKIAA0068; mKIAA1168; P140SRA-1; P140sra1; PIR121; pl-1; SHYC; SRA-1; SRA1
UniProt ID: (Human) Q7L576, (Human) Q96F07, (Mouse) Q7TMB8, (Mouse) Q5SQX6
Entrez Gene ID: (Human) 23191, (Human) 26999, (Mouse) 20430, (Mouse) 76884
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.