Invitrogen
Manufacturer Code:PA528125
Catalog # PIPA528125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 223 and 286 of Human Cytochrome b reductase 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome b reductase 1; cytochrome b561 family, member A2; DCYTB FRRS3 duodenal cytochrome b ferric-chelate reductase 3; Duodenal cytochrome b; Ferric-chelate reductase 3; Plasma membrane ascorbate-dependent reductase CYBRD1
Gene Aliases: CYB561A2; CYBRD1; DCYTB; FRRS3
UniProt ID: (Human) Q53TN4
Entrez Gene ID: (Human) 79901
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.