Invitrogen
Manufacturer Code:PA575653
Catalog # PIPA575653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human CYBA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytochrome b light chain; Cytochrome b(558) alpha chain; cytochrome b(558) alpha-subunit; cytochrome b, alpha polypeptide; Cytochrome b-245 light chain; Cytochrome b-245 light chain Cytochrome b(558) alpha chain Cytochrome b558 subunit alpha Neutrophil cytochrome b 22 kDa polypeptide Superoxide-generating NADPH oxidase light chain subunit p22 phagocyte B-cytochrome p22-phox p22phox CYBA; cytochrome b-245, alpha polypeptide; cytochrome b558 alpha-subunit; Cytochrome b558 subunit alpha; cytochrome beta-558; flavocytochrome b-558 alpha polypeptide; Neutrophil cytochrome b 22 kDa polypeptide; p22 phagocyte B-cytochrome; p22 phox; p22-phox; p22phox; Superoxide-generating NADPH oxidase light chain subunit
Gene Aliases: b558; CYBA; nmf333; p22-PHOX; p22phox
UniProt ID: (Human) P13498, (Mouse) Q61462, (Rat) Q62737
Entrez Gene ID: (Human) 1535, (Mouse) 13057, (Rat) 79129
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.