Invitrogen
Manufacturer Code:MA525143
Catalog # PIMA525143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E8 |
Immunogen | Full length human recombit protein of CYV5R3 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B5R; Cyb5r3 cytochrome b5 reductase 3 0610016L08Rik 2500002N19Rik C85115 Dia-1 Dia1 WU:AL591952.1-001 WU:AL591952.1-002 WU:AL591952.1-003 WU:Cyb5r3 NADH-cytochrome b5 reductase diaphorase 1 diaphorase 1 (NADH); Diaphorase-1; mutant NADH-cytochrome b5 reductase; NADH-cytochrome b5 reductase 3; NADH-cytochrome b5 reductase 3 membrane-bound form; NADH-cytochrome b5 reductase 3 soluble form
Gene Aliases: B5R; CYB5R3; DIA1
UniProt ID: (Human) Q9UEN4
Entrez Gene ID: (Human) 1727
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.