Invitrogen
Manufacturer Code:PA519196
Catalog # PIPA519196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PNLERVGHPKERC) corresponding to the C-terminus amino acids of Cyb5r3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B5R; Cyb5r3 cytochrome b5 reductase 3 0610016L08Rik 2500002N19Rik C85115 Dia-1 Dia1 WU:AL591952.1-001 WU:AL591952.1-002 WU:AL591952.1-003 WU:Cyb5r3 NADH-cytochrome b5 reductase diaphorase 1 diaphorase 1 (NADH); diaphorase 1 (NADH); Diaphorase-1; NADH-cytochrome b5 reductase 3; NADH-cytochrome b5 reductase 3 membrane-bound form; NADH-cytochrome b5 reductase 3 soluble form
Gene Aliases: 0610016L08Rik; 2500002N19Rik; C85115; Cyb5r3; Dia-1; Dia1; WU:AL591952.1-001; WU:AL591952.1-002; WU:AL591952.1-003; WU:Cyb5r3
UniProt ID: (Mouse) Q9DCN2
Entrez Gene ID: (Mouse) 109754
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.