Invitrogen
Manufacturer Code:PA5103405
Catalog # PIPA5103405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Cytochrome b561 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD158F CD158F1 CYB561 CYB561A1 cytochrome b561 Cytochrome b-561 cytochrome b561 family member A1 ferric-chelate reductase 2 FRRS2 KIR2DL5 Mcyt; CGCytb; Chromaffin granule Cyt b561; Cytochrome b-561; Cytochrome b561; cytochrome b561 family, member A1; ferric-chelate reductase 2; Transmembrane ascorbate-dependent reductase CYB561
Gene Aliases: CYB561; CYB561A1; FRRS2; Mcyt
UniProt ID: (Human) P49447, (Mouse) Q60720
Entrez Gene ID: (Human) 1534, (Mouse) 13056
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.