R&D Systems
Manufacturer Code:FAB4227A025
Catalog # FAB4227A025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11G8 |
Immunogen | Human CXCR7 encoding plasmid Accession # P25106 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: ACKR3 chemokine (C-X-C motif) receptor 7 Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7 CMKOR1 CXCR7 CXC-R7 CXCR-7 G protein-coupled receptor GPR159 GPR159G-protein coupled receptor 159 RDC-1 RDC1G-protein coupled receptor RDC1 homolog; Atypical chemokine receptor 3; C-X-C chemokine receptor type 7; chemokine (C-X-C motif) receptor 7; Chemokine orphan receptor 1; CXC-R7; G protein-coupled receptor; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog; RDC-1
Gene Aliases: ACKR3; CMKOR1; CXC-R7; CXCR-7; CXCR7; GPR159; RDC-1; RDC1
UniProt ID: (Human) P25106
Entrez Gene ID: (Human) 57007
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.