Proteintech
Manufacturer Code:148401AP150UL
Catalog # 50-172-9768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Atypical chemokine receptor 3; C-X-C chemokine receptor type 7; chemokine (C-X-C motif) receptor 7; Chemokine orphan receptor 1; Chemokine orphan receptor 1 CMKOR1 CXC R7 CXCR 7 CXCR7 G protein coupled receptor 159 GPR159 RDC 1 RDC1; CXC-R7; CXCR-7; G protein-coupled receptor; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog; RDC-1
Gene Aliases: ACKR3; AW541270; CMKOR1; CXC-R7; CXCR-7; CXCR7; GPR159; RDC-1; RDC1
UniProt ID: (Human) P25106, (Rat) O89039, (Mouse) P56485
Entrez Gene ID: (Human) 57007, (Rat) 84348, (Mouse) 12778
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.