Invitrogen
Manufacturer Code:700995
Catalog # PI700995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 6H35L3 |
Immunogen | proprietary |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Burkitt lymphoma receptor 1; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C-X-C chemokine receptor type 5; CD185; CD185 antigen CXCR-5 CXC-R5 GPR9 MDR15 Monocyte-derived receptor 15 Blr-1 BLR1 Burkitt lymphoma receptor 1 Burkitt&primes lymphoma receptor 1 MDR-15; chemokine (C-X-C motif) receptor 5; CXC-R5; CXCR-5; MDR-15; Monocyte-derived receptor 15
Gene Aliases: BLR1; CD185; CXCR5; MDR15
UniProt ID: (Human) P32302
Entrez Gene ID: (Human) 643
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.