R&D Systems
Manufacturer Code:FAB190N025
Catalog # FAB190N025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 51505 |
Immunogen | NS0 mouse myeloma cell line transfected with human CXCR5 Met1-Phe372 Accession # P32302 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 °C as supplied. |
Protein Aliases: BLR1 BLR1MDR-15 Burkitt lymphoma receptor 1 Burkitt lymphoma receptor 1 GTP binding protein (chemokine (C-X-C motif)receptor 5) Burkitt lymphoma receptor 1 GTP-binding protein CD185 CD185 antigen chemokine (C-X-C motif) receptor 5 C-X-C chemokine receptor type 5 CXCR5 CXC-R5 MDR15CXCR-5 MGC117347 Monocyte-derived receptor 15; Burkitt lymphoma receptor 1; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C-X-C chemokine receptor type 5; CD185; chemokine (C-X-C motif) receptor 5; CXC-R5; CXCR-5; MDR-15; Monocyte-derived receptor 15
Gene Aliases: BLR1; CD185; CXCR5; MDR15
UniProt ID: (Human) P32302
Entrez Gene ID: (Human) 643
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.