R&D Systems
Manufacturer Code:AF6198SP
Catalog # AF6198SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse CXCR5 Met1-Pro57 Glu112-Thr126 Lys191-Phe221 and Leu283-Ile306 joined by linker peptides Accession # Q04683 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BLR1 BLR1MDR-15 Burkitt lymphoma receptor 1 Burkitt lymphoma receptor 1 GTP binding protein (chemokine (C-X-C motif)receptor 5) Burkitt lymphoma receptor 1 GTP-binding protein CD185 CD185 antigen chemokine (C-X-C motif) receptor 5 C-X-C chemokine receptor type 5 CXCR5 CXC-R5 MDR15CXCR-5 MGC117347 Monocyte-derived receptor 15; Burkitt lymphoma receptor 1; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C-X-C chemokine receptor type 5; CD185; chemokine (C-X-C motif) receptor 5; CXC-R5; CXCR-5; MDR-15; Monocyte-derived receptor 15
Gene Aliases: BLR1; CD185; CXCR5; MDR15
UniProt ID: (Human) P32302
Entrez Gene ID: (Human) 643
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.