R&D Systems
Manufacturer Code:MAB21651RSP
Catalog # MAB21651R-SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 247506R |
Immunogen | Y3 rat myeloid cell line transfected with mouse CXCR4 Met1-Ser359 Accession # P70658 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
Protein Aliases: C-X-C chemokine receptor type 4; CD184; CD184 antigen; CD184 chemokine (C-X-C motif) receptor 4 chemokine (C-X-C motif) receptor 4 (fusin) C-X-C chemokine receptor type 4 CXC-R4 CXCR-4 D2S201E D2S201ESDF-1 receptor FB22 Fusin HM89 HM89NPYRL HSY3RR LAP3 LAP-3 LCR1 LESTR LESTRCD184 antigen Leukocyte-derived seven transmembrane domain receptor leukocyte-derived seven-transmembrane-domain receptor lipopolysaccharide-associated protein 3 neuropeptide Y receptor Y3 NPY3R NPY3RWHIM NPYR NPYRL NPYY3R pCXCR4 seven transmembrane helix receptor seven-transmembrane-segment receptor spleen Stromal cell-derived factor 1 receptor WHIMS; chemokine (C-X-C motif) receptor 4; chemokine receptor; CXC-R4; FB22; Fusin; HM89; LAP-3; LCR1; LESTR; Leukocyte-derived seven transmembrane domain receptor; Lipopolysaccharide-associated protein 3; LPS-associated protein 3; neuropeptide Y receptor Y3; neuropeptide Y3 receptor; NPYRL; SDF-1 receptor; seven transmembrane helix receptor; seven-transmembrane-segment receptor, spleen; Stromal cell-derived factor 1 receptor
Gene Aliases: CD184; CXCR4; D2S201E; FB22; HM89; HSY3RR; LAP-3; LAP3; LCR1; LESTR; NPY3R; NPYR; NPYRL; NPYY3R; WHIM; WHIMS
UniProt ID: (Human) P61073
Entrez Gene ID: (Human) 7852
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.