Novus Biologicals
Manufacturer Code:NB1001631
Catalog # NB1001631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 48311.211 |
Immunogen | Human CXCR2 transfected NSO mouse myeloma cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-X-C chemokine receptor type 2; CD182; CD182 antigen CDw128b chemokine (C-X-C motif) receptor 2 chemokine (CXC) receptor 2 C-X-C chemokine receptor type 2 CXC-R2 CXCR-2 CXCR2 gene for IL8 receptor type B GRO/MGSA receptor High affinity interleukin-8 receptor B IL-8 receptor type 2 IL-8R B IL8R2 IL8RA interleukin 8 receptor B interleukin 8 receptor type 2 interleukin-8 receptor type B; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (CXC) receptor 2; CXC-R2; CXCR-2; CXCR2 gene for IL8 receptor type B; GRO/MGSA receptor; High affinity interleukin-8 receptor B; IL-8 receptor type 2; IL-8R B; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B
Gene Aliases: CD182; CDw128b; CMKAR2; CXCR2; IL8R2; IL8RA; IL8RB
UniProt ID: (Human) P25025
Entrez Gene ID: (Human) 3579
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.