Invitrogen
Manufacturer Code:PA5100951
Catalog # PIPA5100951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CXCR2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C Cmotif chemokine C X C motif chemokine CC motif chemokine CCmotif chemokine CD128 CD182 CD182 antigen CDw128 CDw128b chemokine (C-X-C motif) receptor 2 chemokine (CXC) receptor 2 chemokine (C-X-C) receptor 2 Cmkar2 CXC C-X-C chemokine receptor type 2 CXC motif chemokine C-X-C motif chemokine receptor 2 Cxcr2 CXC-R2 CXCR-2 CXCR2 gene for IL8 receptor type B Gpcr16 GRO/MGSA receptor high affinity interleukin-8 receptor B IL-8 receptor alpha chain Il-8 receptor b IL-8 receptor beta IL-8 receptor type 2 IL-8R B IL8R2 IL8RA Il8rb IL-8rb IL-8Rh interleukin 8 receptor B interleukin 8 receptor type 2 interleukin 8 receptor beta interleukin-8 receptor type B mIL-8RH; C-X-C chemokine receptor type 2; CD182; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (C-X-C) receptor 2; chemokine (CXC) receptor 2; CXC-R2; CXCR-2; CXCR2 gene for IL8 receptor type B; GRO/MGSA receptor; High affinity interleukin-8 receptor B; IL-8 receptor alpha chain; IL-8 receptor type 2; IL-8R B; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B
Gene Aliases: CD128; CD182; CDw128; CDw128b; CMKAR2; CXCR2; Gpcr16; IL-8rb; IL-8Rh; IL8R2; IL8RA; IL8RB; mIL-8RH
UniProt ID: (Human) P25025, (Mouse) P35343
Entrez Gene ID: (Human) 3579, (Mouse) 12765
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.