R&D Systems
Manufacturer Code:MAB8628SP
Catalog # MAB8628SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1122A |
Immunogen | Mouse CXCR1/IL-8 RA Peptide Accession # Q810W6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: C-X-C chemokine receptor type 1; CD128 CD181 CD181 antigen CDw128aC-C chemokine (C-X-C motif) receptor 1 CKR-1 CMKAR1 C-X-C chemokine receptor type 1 CXC-R1 CXCR-1 High affinity interleukin-8 receptor A IL-8 RA IL-8 receptor type 1 IL-8R A IL8R1 IL8RA IL8RAC-C-CKR-1 IL8RBA interleukin 8 receptor alpha interleukin-8 receptor type 1 interleukin-8 receptor type A; CD181; CDw128a; chemokine (C-X-C motif) receptor 1; CXC-R1; CXCR-1; High affinity interleukin-8 receptor A; IL-8 receptor type 1; IL-8R A; interleukin 8 receptor, alpha; interleukin-8 receptor type 1; interleukin-8 receptor type A
Gene Aliases: C-C; C-C-CKR-1; CD128; CD181; CDw128a; CKR-1; CMKAR1; CXCR1; IL8R1; IL8RA; IL8RBA
UniProt ID: (Human) P25024
Entrez Gene ID: (Human) 3577
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.