Novus Biologicals
Manufacturer Code:NLS806
Catalog # NLS806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from N-terminal extracellular domain of human CXCR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X-C chemokine receptor type 1; CD128 CD181 CD181 antigen CDw128aC-C chemokine (C-X-C motif) receptor 1 CKR-1 CMKAR1 C-X-C chemokine receptor type 1 CXC-R1 CXCR-1 High affinity interleukin-8 receptor A IL-8 receptor type 1 IL-8R A IL8R1 IL8RAC-C-CKR-1 IL8RBA interleukin 8 receptor alpha interleukin-8 receptor type 1 interleukin-8 receptor type A; CD181; CDw128a; chemokine (C-X-C motif) receptor 1; CXC-R1; CXCR-1; High affinity interleukin-8 receptor A; IL-8 receptor type 1; IL-8R A; interleukin 8 receptor, alpha; interleukin-8 receptor type 1; interleukin-8 receptor type A
Gene Aliases: C-C; C-C-CKR-1; CD128; CD181; CDw128a; CKR-1; CMKAR1; CXCR1; IL8R1; IL8RA; IL8RBA
UniProt ID: (Human) P25024
Entrez Gene ID: (Human) 3577
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.