R&D Systems
Manufacturer Code:IC392G100UG
Catalog # IC392G100UG
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 49106 |
Immunogen | <EM>E. coli</EM>-derived recombinant human CXCL9/MIG Thr23-Thr125 Accession # Q07325 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 9; chemokine (C-X-C motif) ligand 9; chemokine (C-X-C motif) ligand 9 CMK crg-10 C-X-C motif chemokine 9 Gamma-interferon-induced monokine Humig MIG MIGSmall-inducible cytokine B9 monokine induced by gamma interferon SCYB9Monokine induced by interferon-gamma; Gamma-interferon-induced monokine; HuMIG; monokine induced by gamma interferon; Monokine induced by interferon-gamma; Small-inducible cytokine B9
Gene Aliases: CMK; crg-10; CXCL9; Humig; MIG; SCYB9
UniProt ID: (Human) Q07325
Entrez Gene ID: (Human) 4283
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.