Novus Biologicals
Manufacturer Code:NBP189921
Catalog # NBP189921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GQTKRNLAKGKEESLDSDLYAELRCMCIKTTSGIHPKNIQSLEVIGKGTHCNQVEVIATLKDGRKICLDPDAPRIKKIVQKKLAGDESAD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-TG; Beta-TG connective tissue-activating peptide III CTAP3CTAP-III CTAPIII CXC chemokine ligand 7 C-X-C motif chemokine 7 CXCL7b-TG1 LA-PF4 LDGFTC1 Leukocyte-derived growth factor low-affinity platelet factor IV Macrophage-derived growth factor MDGFTC2 NAP-2 NAP-2-L1 neutrophil-activating peptide 2 neutrophil-activating peptide-2 PBPTHBGB platelet basic protein pro-platelet basic protein (chemokine (C-X-C motif) ligand 7) SCAR10 SCYB7beta-thromboglobulin small inducible cytokine B7 small inducible cytokine subfamily B member 7 Small-inducible cytokine B7 TGB TGB1B-TG1 THBGB1 thrombocidin 1 thrombocidin 2 thromboglobulin beta-1; Beta-thromboglobulin; C-X-C motif chemokine 7; chemokine (C-X-C motif) ligand 7; Connective tissue-activating peptide III; Connective tissue-activating peptide III(1-81); CTAP-III; CTAP-III(1-81); CXC chemokine ligand 7; LA-PF4; LDGF; Leukocyte-derived growth factor; Low-affinity platelet factor IV; Macrophage-derived growth factor; MDGF; NAP-2; NAP-2(1-63); NAP-2(1-66); NAP-2(73); NAP-2(74); Neutrophil-activating peptide 2; Neutrophil-activating peptide 2(1-63); Neutrophil-activating peptide 2(1-66); Neutrophil-activating peptide 2(73); Neutrophil-activating peptide 2(74); neutrophil-activating peptide-2; PBP; Platelet basic protein; pro-platelet basic protein (chemokine (C-X-C motif) ligand 7); small inducible cytokine B7; small inducible cytokine subfamily B, member 7; Small-inducible cytokine B7; TC-1; TC-2; thrombocidin 1; thrombocidin 2; thromboglobulin, beta-1
Gene Aliases: B-TG1; Beta-TG; CTAP-III; CTAP3; CTAPIII; CXCL7; LA-PF4; LDGF; MDGF; NAP-2; PBP; PPBP; SCYB7; TC1; TC2; TGB; TGB1; THBGB; THBGB1
UniProt ID: (Human) P02775
Entrez Gene ID: (Human) 5473
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.