Novus Biologicals
Manufacturer Code:NBP2122310.025MG
Catalog # NB019119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0216-10L19 |
Immunogen | Human CXCL5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 5; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 5 C-X-C motif chemokine 5 ENA-78 ENA-78(1-78) epithelial-derived neutrophil activating protein 78 epithelial-derived neutrophil-activating peptide 78 Epithelial-derived neutrophil-activating protein 78 Neutrophil-activating peptide ENA-78 neutrophil-activating protein 78 SCYB5ENA78 small inducible cytokine B5 small inducible cytokine subfamily B (Cys-X-Cys) member 5 (epithelial-derivedneutrophil-activating peptide 78) Small-inducible cytokine B5; ENA-78(1-78); ENA-78(8-78); ENA-78(9-78); Epithelial-derived neutrophil-activating protein 78; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derived neutrophil-activating peptide 78); Small-inducible cytokine B5
Gene Aliases: CXCL5; ENA-78; ENA78; SCYB5
UniProt ID: (Human) P42830
Entrez Gene ID: (Human) 6374
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.