Invitrogen
Manufacturer Code:PA579112
Catalog # PIPA579112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat CXCL5 recombit protein (Position: A38-Q130). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 5; C-X-C motif chemokine 5 Cytokine LIX Small-inducible cytokine B5 Cxcl5 Scyb5; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); CXC chemokine LIX; Cytokine LIX; Small-inducible cytokine B5
Gene Aliases: Cxcl5; Scyb5
UniProt ID: (Rat) P97885
Entrez Gene ID: (Rat) 60665
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.