Novus Biologicals
Manufacturer Code:NBP212225G
Catalog # NBP212225G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0206-17P3 |
Immunogen | Mouse CXCL14 |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: BMAC BRAKKEC chemokine (C-X-C motif) ligand 14 Chemokine BRAK member 14 (BRAK) MIP-2 gamma MIP2G MIP-2G NJACKec SCYB14MGC10687 Small-inducible cytokine B14; bolekine; breast and kidney; C-X-C motif chemokine 14; chemokine (C-X-C motif) ligand 14; Chemokine BRAK; CXC chemokine in breast and kidney; MIP-2 gamma; MIP-2G; small inducible cytokine subfamily B (Cys-X-Cys), member 14 (BRAK); Small-inducible cytokine B14; tumor-suppressing chemokine
Gene Aliases: BMAC; BRAK; CXCL14; KEC; KS1; MIP-2g; MIP2G; NJAC; PSEC0212; SCYB14; UNQ240/PRO273
UniProt ID: (Human) O95715
Entrez Gene ID: (Human) 9547
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.