Invitrogen
Manufacturer Code:PA546815
Catalog # PIPA546815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CXCL14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 14; Chemokine (C-X-C motif) ligand 14 BMAC BRAK KEC KS1 MGC10687 MIP-2g MIP2G NJAC SCYB14; small inducible cytokine B14
Gene Aliases: BRAK
Entrez Gene ID: (Bovine) 511771, (Horse) 100072697, (Dog) 610078, (Guinea pig) 100716180, (Rat) 306748
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.