Invitrogen
Manufacturer Code:PA528820
Catalog # PIPA528820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 47 and 111 of Human BRAK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell and monocyte-activating chemokine; BMAC BRAK KEC KS1 MIP-2g MIP2G NJAC SCYB14 C-X-C motif chemokine 14 CXC chemokine in breast and kidney MIP-2 gamma bolekine breast and kidney chemokine BRAK small inducible cytokine subfamily B (Cys-X-Cys) member 14 (BRAK) small-inducible cytokine B14 tumor-suppressing chemokine; bolekine; breast and kidney; C-X-C motif chemokine 14; chemokine (C-X-C motif) ligand 14; Chemokine BRAK; CXC chemokine in breast and kidney; Kidney-expressed chemokine CXC; MIP-2 gamma; MIP-2G; musculus CXC chemokine MIP-2gamma; small inducible cytokine subfamily B (Cys-X-Cys), member 14; small inducible cytokine subfamily B (Cys-X-Cys), member 14 (BRAK); Small-inducible cytokine B14; tumor-suppressing chemokine
Gene Aliases: 1110031L23Rik; 1200006I23Rik; AI414372; BMAC; bolekine; BRAK; CXCL14; KEC; KS1; MIP-2g; MIP2G; MIP2gamma; NJAC; PSEC0212; SCYB14; UNQ240/PRO273
UniProt ID: (Human) O95715, (Mouse) Q9WUQ5
Entrez Gene ID: (Human) 9547, (Mouse) 57266
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.