R&D Systems
Manufacturer Code:MAB470RSP
Catalog # MAB470RSP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 143614R |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse CXCL13 Ile22-Ala109 Accession # O55038 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Angie; ANGIE ANGIE2 B cell-attracting chemokine 1 B lymphocyte chemoattractant BCA1 BCA-1 BCA1B-cell chemoattractant BCA-1CXC chemokine BLC B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1) BLC BLCSmall-inducible cytokine B13 BLR1L B; B cell-attracting chemokine 1; B lymphocyte chemoattractant; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); B-lymphocyte chemoattractant; BCA-1; C-X-C motif chemokine 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC chemokine BLC; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); Small-inducible cytokine B13
Gene Aliases: ANGIE; ANGIE2; BCA-1; BCA1; BLC; BLR1L; CXCL13; SCYB13
UniProt ID: (Human) O43927
Entrez Gene ID: (Human) 10563
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.