Novus Biologicals
Manufacturer Code:NBP216041SS
Catalog # NBP216041SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to a C-terminal region of human CXCL13 (between amino acids 50-109) [UniProt O43927] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: Angie; ANGIE ANGIE2 B cell-attracting chemokine 1 B lymphocyte chemoattractant BCA1B-cell chemoattractant BCA-1CXC chemokine BLC B-cell-homing chemokine (ligand for Burkitt&primes lymphoma receptor-1) BLCSmall-inducible cytokine B13 BLR1L B-lymphocyte chemoattractant chemokine (C-X-C motif) ligand 13 chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant) C-X-C motif chemokine 13 SCYB13B-cell-attracting chemokine 1 small inducible cytokine B subfamily (Cys-X-Cys motif) member 13 (B-cellchemoattractant); B cell-attracting chemokine 1; B lymphocyte chemoattractant; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); B-lymphocyte chemoattractant; BCA-1; C-X-C motif chemokine 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC chemokine BLC; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); Small-inducible cytokine B13
Gene Aliases: ANGIE; ANGIE2; BCA-1; BCA1; BLC; BLR1L; CXCL13; SCYB13
UniProt ID: (Human) O43927
Entrez Gene ID: (Human) 10563
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.