Invitrogen
Manufacturer Code:MA523666
Catalog # PIMA523666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 53610 |
Immunogen | E. coli-derived recombit human CXCL13/BLC/BCA-1 Val23-Arg94 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Angie; ANGIE ANGIE2 BCA-1 BCA1 BLC BLR1L SCYB13 B-cell chemoattractant B-cell-attracting chemokine 1 B-cell-homing chemokine (ligand for Burkitt&primes lymphoma receptor-1) B-lymphocyte chemoattractant C-X-C motif chemokine 13 CXC chemokine BLC b cell-attracting chemokine 1 b lymphocyte chemoattractant chem; B cell-attracting chemokine 1; B lymphocyte chemoattractant; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); B-lymphocyte chemoattractant; BCA-1; C-X-C motif chemokine 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC chemokine BLC; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); Small-inducible cytokine B13
Gene Aliases: ANGIE; ANGIE2; BCA-1; BCA1; BLC; BLR1L; CXCL13; SCYB13
UniProt ID: (Human) O43927
Entrez Gene ID: (Human) 10563
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.