Invitrogen
Manufacturer Code:MA523871
Catalog # PIMA523871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 143614 |
Immunogen | E. coli-derived recombit mouse CXCL13 Ile22-Ala109 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ANGIE ANGIE2 BCA-1 BCA1 BLC BLR1L SCYB13 B-cell chemoattractant B-cell-attracting chemokine 1 B-cell-homing chemokine (ligand for Burkitt&primes lymphoma receptor-1) B-lymphocyte chemoattractant C-X-C motif chemokine 13 CXC chemokine BLC b cell-attracting chemokine 1 b lymphocyte chemoattractant chem; B lymphocyte chemoattractant; C-X-C motif chemokine 13; CXC chemokine BLC; small inducible cytokine subfamily B (Cys-X-Cys), member 13; Small-inducible cytokine B13
Gene Aliases: 4631412M08Rik; Angie; ANGIE2; BCA-1; BLC; BLR1L; Cxcl13; Scyb13
UniProt ID: (Mouse) O55038
Entrez Gene ID: (Mouse) 55985
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.