Novus Biologicals
Manufacturer Code:NBP2294800.05MG
Catalog # NBP22948005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antisera to human SDF 1 alpha were raised by repeated immunisations of rabbits with highly purified antigen. Purified IgG prepared by affinity chromatography. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: C-X-C motif chemokine 12; chemokine (C-X-C motif) ligand 12; CXCL12 chemokine (C-X-C motif) ligand 12 IRH PBSF SCYB12 SDF1 SDF1A SDF1B TLSF TPAR1; Intercrine reduced in hepatomas; IRH; PBSF; Pre-B cell growth-stimulating factor; SDF-1; SDF-1-alpha(3-67); SDF-1-beta(3-72); Stromal cell-derived factor 1
Gene Aliases: CXCL12; IRH; PBSF; SCYB12; SDF1; SDF1A; SDF1B; TLSF; TPAR1
UniProt ID: (Human) P48061
Entrez Gene ID: (Human) 6387
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.