Novus Biologicals
Manufacturer Code:NBP2122210.025MG
Catalog # NB019110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0211-9N26 |
Immunogen | Human SDF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 12; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 C-X-C motif chemokine 12 hIRH hSDF-1 Intercrine reduced in hepatomas IRH PBSFSDF-1a Pre-B cell growth-stimulating factor SCYB12 SDF-1 SDF1A SDF1B SDF1SDF-1b stromal cell-derived factor 1 TLSF TLSF-a TLSF-b TPAR1; Intercrine reduced in hepatomas; IRH; PBSF; Pre-B cell growth-stimulating factor; SDF-1; SDF-1-alpha(3-67); SDF-1-beta(3-72); Stromal cell-derived factor 1
Gene Aliases: CXCL12; IRH; PBSF; SCYB12; SDF1; SDF1A; SDF1B; TLSF; TPAR1
UniProt ID: (Human) P48061
Entrez Gene ID: (Human) 6387
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.