R&D Systems
Manufacturer Code:MAB310500
Catalog # MAB310500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 79014 |
Immunogen | <EM>E. coli</EM>-derived recombinant human CXCL12/SDF-1 alpha Lys22-Lys89 Accession # Q6ICW0 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 12; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 C-X-C motif chemokine 12 hIRH hSDF-1 Intercrine reduced in hepatomas IRH PBSF PBSFSDF-1a Pre-B cell growth-stimulating factor SCYB12 SDF1 SDF-1 SDF1A SDF1B SDF1SDF-1b stromal cell-derived factor 1 TLSF TLSF-a TLSF-b TPAR1; Intercrine reduced in hepatomas; IRH; PBSF; Pre-B cell growth-stimulating factor; SDF-1; SDF-1-alpha(3-67); SDF-1-beta(3-72); Stromal cell-derived factor 1
Gene Aliases: CXCL12; IRH; PBSF; SCYB12; SDF1; SDF1A; SDF1B; TLSF; TPAR1
UniProt ID: (Human) P48061
Entrez Gene ID: (Human) 6387
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.