Invitrogen
Manufacturer Code:PA595549
Catalog # PIPA595549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human CXCL12 recombit protein (Position: V24-M93). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C-X-C motif chemokine 12; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); Intercrine reduced in hepatomas; IRH; PBSF; Pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SDF-1; SDF-1 gamma; SDF-1-alpha(3-67); SDF-1-beta(3-72); Stromal cell-derived factor 1; Stromal cell-derived factor 1 SDF-1 hSDF-1 C-X-C motif chemokine 12 Intercrine reduced in hepatomas IRH hIRH Pre-B cell growth-stimulating factor PBSF SDF-1-beta(3-72) SDF-1-alpha(3-67) CXCL12 SDF1 SDF1A SDF1B; stromal cell-derived factor-1 gamma; Thymic lymphoma cell-stimulating factor; TLSF; TPAR1
Gene Aliases: CXCL12; IRH; PBSF; SCYB12; SDF1; SDF1A; SDF1B; TLSF; TPAR1
UniProt ID: (Human) P48061, (Mouse) P40224
Entrez Gene ID: (Human) 6387, (Rat) 24772, (Mouse) 20315
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.