Invitrogen
Manufacturer Code:PA547767
Catalog # PIPA547767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse CXCL11/I-TAC Phe22-Met100 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 11; CXCL11 H174 I-TAC IP-9 IP9 MGC102770 SCYB11 SCYB9B b-R1 C-X-C motif chemokine 11 Small-inducible cytokine B11 Interferon-inducible T-cell alpha chemoattractant Interferon-gamma-inducible protein 9 Beta-R1; I-TAC; Interferon-inducible T-cell alpha chemoattractant; small inducible cytokine subfamily B (Cys-X-Cys), member 11; Small-inducible cytokine B11
Gene Aliases: b-R1; betaR1; Cxc11; Cxcl11; H174; I-tac; Ip9; Itac; Scyb11; Scyb9b
UniProt ID: (Mouse) Q9JHH5
Entrez Gene ID: (Mouse) 56066
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.