Invitrogen
Manufacturer Code:PA5116353
Catalog # PIPA5116353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | recombinant protein of human CXCL11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-R1; betaR1 Beta-R1 b-R1 C Cmotif chemokine C X C motif chemokine CC motif chemokine CCmotif chemokine chemokine (C-X-C motif) ligand 11 CXC CXC motif chemokine C-X-C motif chemokine 11 C-X-C motif chemokine ligand 11 Cxc11 CXCL Cxcl11 H174 Interferon gamma-inducible protein 9 interferon-inducible T-cell alpha chemoattractant Ip9 IP-9 ITAC I-TAC MGC102770 Scyb11 SCYB9B small inducible cytokine B11 small inducible cytokine subfamily B (Cys-X-Cys) member 11 small inducible cytokine subfamily B (Cys-X-Cys) member 9B small-inducible cytokine B11; C-X-C motif chemokine 11; chemokine (C-X-C motif) ligand 11; H174; I-TAC; Interferon gamma-inducible protein 9; Interferon-inducible T-cell alpha chemoattractant; IP-9; small inducible cytokine B11; small inducible cytokine subfamily B (Cys-X-Cys), member 11; small inducible cytokine subfamily B (Cys-X-Cys), member 9B; Small-inducible cytokine B11
Gene Aliases: b-R1; CXCL11; H174; I-TAC; IP-9; IP9; ITAC; SCYB11; SCYB9B
UniProt ID: (Human) O14625
Entrez Gene ID: (Human) 6373, (Rat) 305236
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.