R&D Systems
Manufacturer Code:IC266RT100UG
Catalog # IC266RT100UG
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 33036R |
Immunogen | <i>E. coli</i>-derived recombinant human CXCL10/IP-10/CRG-2 Val22-Pro98 Accession &num P02778.2 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 10 kDa interferon gamma-induced protein; C-X-C motif chemokine 10; C7 chemokine (C-X-C motif) ligand 10 crg-2 CRG2 gIP-10 IFI10 INP10 IP-10 mob-1 SCYB10; chemokine (C-X-C motif) ligand 10; CXCL10(1-73); gamma IP10; Gamma-IP10; interferon-inducible cytokine IP-10; protein 10 from interferon (gamma)-induced cell line; small inducible cytokine subfamily B (Cys-X-Cys), member 10; Small-inducible cytokine B10
Gene Aliases: C7; crg-2; CXCL10; gIP-10; IFI10; INP10; IP-10; mob-1; SCYB10
UniProt ID: (Human) P02778
Entrez Gene ID: (Human) 3627
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.