Invitrogen
Manufacturer Code:701225
Catalog # PI701225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 10H11L3 |
Immunogen | proprietary |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 10 kDa interferon gamma-induced protein; C-X-C motif chemokine 10; C7; Gamma-IP10; interferon activated gene 10; Interferon Gamma Inducible Protein 10 C7 CRG-2 INP10 IP-10 IP10 Ifi10 Scyb10 gIP-10 mob-1; Interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; small inducible cytokine B subfamily (Cys-X-Cys), member 10; Small-inducible cytokine B10
Gene Aliases: C7; CRG-2; Crg2; Cxcl10; gIP-10; Ifi10; INP10; IP-10; IP10; mob-1; Scyb10
UniProt ID: (Mouse) P17515
Entrez Gene ID: (Mouse) 15945
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.