Invitrogen
Manufacturer Code:PA586508
Catalog # PIPA586508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 41-90 of Human GROalpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-chemokine; C-X-C motif chemokine 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); CINC-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; gro; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); Growth-regulated alpha protein; Growth-regulated alpha protein C-X-C motif chemokine 1 GRO-alpha(1-73) Melanoma growth stimulatory activity MGSA Neutrophil-activating protein 3 NAP-3 GRO GRO1 GROA MGSA SCYB1 CXCL1 GRO alpha; Hematopoietic synergistic factor; HSF; KC(5-72); KC-T; KC/GR)-alpha; KC/GRO-alpha; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; NAP-3; Neutrophil-activating protein 3; Platelet-derived growth factor-inducible protein KC; Secretory protein N51
Gene Aliases: CINC-1; Cinc1; CXCL1; FSP; GRO; GRO1; GROA; KC; MGSA; MGSA-a; N51; NAP-3; SCYB1
UniProt ID: (Human) P09341, (Rat) P14095, (Mouse) P12850
Entrez Gene ID: (Human) 2919, (Rat) 81503, (Mouse) 14825
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.