Invitrogen
Manufacturer Code:PA129220
Catalog # PIPA129220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment - amino acids 1-72 of rat KC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-chemokine; C-X-C motif chemokine 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); CINC-1; Cytokine-induced neutrophil chemoattractant 1; Fsp Gro1 KC Mgsa N51 Scyb1 gro; gro; GRO1 oncogene; Growth-regulated alpha protein; Hematopoietic synergistic factor; HSF; KC(5-72); KC-T; KC/GR)-alpha; KC/GRO-alpha; Platelet-derived growth factor-inducible protein KC; Secretory protein N51
Gene Aliases: CINC-1; Cinc1; Cxcl1; Fsp; Gro; Gro1; KC; Mgsa; N51; Scyb1
UniProt ID: (Rat) P14095, (Mouse) P12850
Entrez Gene ID: (Rat) 81503, (Mouse) 14825
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.