Novus Biologicals
Manufacturer Code:NBP290355100UL
Catalog # NB022163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombinant Mouse CXADR protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 46 kD coxsackievirus and adenovirus receptor (CAR) protein; CAR; CAR10Coxsackievirus B-adenovirus receptor CAR4/6 coxsackie virus and adenovirus receptor coxsackie virus B receptor coxsackievirus and adenovirus receptor CVB3 binding protein CVB3-binding protein hCAR HCVADR; Coxsackievirus and adenovirus receptor; Coxsackievirus B-adenovirus receptor; CVB3-binding protein; HCVADR
Gene Aliases: CAR; CAR4/6; CXADR; HCAR
UniProt ID: (Human) P78310
Entrez Gene ID: (Human) 1525
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.