R&D Systems
Manufacturer Code:BAF2654
Catalog # BAF2654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CXADR Leu20-Gly237 Accession # P97792 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 46 kD coxsackievirus and adenovirus receptor (CAR) protein; CAR; CAR10Coxsackievirus B-adenovirus receptor CAR4/6 coxsackie virus and adenovirus receptor coxsackie virus B receptor coxsackievirus and adenovirus receptor CVB3 binding protein CVB3 BP CVB3-binding protein hCAR HCVADR; Coxsackievirus and adenovirus receptor; Coxsackievirus B-adenovirus receptor; CVB3-binding protein; HCVADR
Gene Aliases: CAR; CAR4/6; CXADR; HCAR
UniProt ID: (Human) P78310
Entrez Gene ID: (Human) 1525
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.