Proteintech
Manufacturer Code:117771AP150UL
Catalog # 50-172-7447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CXADR Fusion Protein Ag2369 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 46 kD coxsackievirus and adenovirus receptor (CAR) protein; CAR; CAR CVB3 binding protein CXADR HCAR HCVADR; Coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; Coxsackievirus B-adenovirus receptor; CVB3-binding protein; HCVADR; rCAR
Gene Aliases: 2610206D03Rik; AU016810; AW553441; CAR; Car1; CAR4/6; CXADR; HCAR; MCAR; MCVADR
UniProt ID: (Human) P78310, (Mouse) P97792, (Rat) Q9R066
Entrez Gene ID: (Human) 1525, (Mouse) 13052, (Rat) 89843
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.