R&D Systems
Manufacturer Code:FAB5825A025
Catalog # FAB5825A025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse CX3CR1 Met1-Thr32 Leu92-Lys104 Thr169-Val196 Lys258-Leu274 Accession # Q9Z0D9 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Goat Polyclonal Antibody
Protein Aliases: Beta chemokine receptor-like 1; Beta chemokine receptor-like 1 CCRL1 chemokine (C-C) receptor-like 1 chemokine (C-X3-C motif) receptor 1 chemokine (C-X3-C) receptor 1 CMKbRL1 CMK-BRL1 CMK-BRL-1 CMKDR1 CX3C chemokine receptor 1 Fractalkine receptor G protein-coupled receptor 13 GPR13G-protein coupled receptor 13 GPRV28 V28 V28C-X3-C CKR-1; C-X3-C CKR-1; chemokine (C-C) receptor-like 1; chemokine (C-X3-C motif) receptor 1; chemokine (C-X3-C) receptor 1; CMK-BRL-1; CMK-BRL1; CX3C chemokine receptor 1; Fractalkine receptor; G protein-coupled receptor 13; G-protein coupled receptor 13; V28
Gene Aliases: CCRL1; CMKBRL1; CMKDR1; CX3CR1; GPR13; GPRV28; V28
UniProt ID: (Human) P49238
Entrez Gene ID: (Human) 1524
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.