R&D Systems
Manufacturer Code:IC3652RS100UG
Catalog # IC3652RS100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 81506R |
Immunogen | NS0-derived and <i>E. coli</i>-derived Recombinant human CX3CL1/Fracktalkine Gln25-Arg339 Accession &num P78423 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABCD-3 C3Xkine chemokine (C-X3-C motif) ligand 1 CX3C membrane-anchored chemokine CXC3 CXC3C FKN fractalkine Neurotactin neurotactin) NTNsmall inducible cytokine subfamily D (Cys-X3-Cys) member 1 (fractalkine NTTSmall-inducible cytokine D1 SCYD1C-X3-C motif chemokine 1 small inducible cytokine subfamily D (Cys-X3-Cys) member-1; C-X3-C motif chemokine 1; chemokine (C-X3-C motif) ligand 1; CX3C membrane-anchored chemokine; Fractalkine; Neurotactin; Processed fractalkine; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; Small-inducible cytokine D1
Gene Aliases: A-152E5.2; ABCD-3; C3Xkine; CX3CL1; CXC3; CXC3C; FKN; fractalkine; neurotactin; NTN; NTT; SCYD1
UniProt ID: (Human) P78423
Entrez Gene ID: (Human) 6376
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.