Invitrogen
Manufacturer Code:PA547728
Catalog # PIPA547728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit rat CX3CL1/Fractalkine Gln25-Gly100 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X3-C motif chemokine 1; CX3C membrane-anchored chemokine; CX3CL1 FKN NTT SCYD1 C-X3-C motif chemokine 1 CX3C membrane-anchored chemokine Neurotactin Small-inducible cytokine D1; Fractalkine; Neurotactin; Processed fractalkine; small inducible cytokine subfamily D, 1; Small-inducible cytokine D1
Gene Aliases: Acc1; Cx3c; Cx3cl1; Fkn; Scyd1
UniProt ID: (Rat) O55145
Entrez Gene ID: (Rat) 89808
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.