OriGene
Manufacturer Code:TA808677
Catalog # 50-168-2292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 145-472 of human CWC27 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-10; CWC27 spliceosome-associated protein homolog; NY-CO-10 RPSKA SDCCAG-10 SDCCAG10; peptidyl-prolyl cis-trans isomerase SDCCAG10; PPIase CWC27; PPIase SDCCAG10; Probable inactive peptidyl-prolyl cis-trans isomerase CWC27 homolog; Serologically defined colon cancer antigen 10; Spliceosome-associated protein CWC27 homolog
Gene Aliases: CWC27; NY-CO-10; SDCCAG-10; SDCCAG10; UNQ438/PRO871
UniProt ID: (Human) Q6UX04
Entrez Gene ID: (Human) 10283
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.