Novus Biologicals
Manufacturer Code:NBP156858
Catalog # NBP156858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CUTC(cutC copper transporter homolog (E. coli)) The peptide sequence was selected from the middle region of CUTC. Peptide sequence LEGLPLIKRLIEQAKGRIVVMPGGGITDRNLQRILEGSGATEFHCSARST. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI-32 copper homeostasis protein cutC homolog cutC copper transporter homolog (E. coli) RP11-483F11.3; Copper homeostasis protein cutC homolog; cutC copper transporter homolog
Gene Aliases: CGI-32; CUTC
UniProt ID: (Human) Q9NTM9
Entrez Gene ID: (Human) 51076
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.